Lead in powdered paint, certified reference material, Supelco®
Lieferant: Merck
SQC074-30G
SQC074-30GEA
107
EUR
SQC074-30G
Lead in powdered paint, certified reference material, Supelco®
Standards
Kationischer Standards
Blei-Standards
ICP and AAS suitable.
Values of analyte vary lot to lot.
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.