p-Anisaldehyd-Lösung, Sprühreagenz, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
Anisaldehyd
SRA1-1KT
SRA1-1KTEA
142
EUR
SRA1-1KT
p-Anisaldehyd-Lösung, Sprühreagenz, Sigma-Aldrich®
p-Anisaldehyd
Excellent reagent for detecting a wide variety of natural products on TLC plates: steroids, prostaglandins, carbohydrates, phenols, glycosides, sapogenins, antibiotics, mycotoxins, antioxidants etc. A range of colors is produced on heating, adding a qualitative aspect to the detection process.
Formel:
C₈H₈O₂ Molecular Weight: 136,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003385 CAS-Nummer: 123-11-5 ADR: 9,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...