Twin-column adsorption apparatus
Lieferant: STANHOPE SETA
14060-0
STAN14060-0EA 0 EUR
STAN14060-0
Twin-column adsorption apparatus
Apparate zur Mineralöluntersuchung
The twin-column adsorption apparatus can measures fluorescent indicator adsorption in liquid petroleum products.
  • Determines volume % of saturates, olefins and aromatics in petroleum fractions
  • Analyses two samples simultaneously
  • Integral ultra violet illumination