Freezing point apparatus
Lieferant: STANHOPE SETA
16990-2
STAN16990-2EA 0 EUR
STAN16990-2
Freezing point apparatus
Schmelzpunkt-Bestimmungsgeräte
The freezing point apparatus detects solid hydrocarbon crystals and uses reference method.
Standards: ASTM D2386; IP 16; ISO 3013; NF M07-048; FTMS 791 1411; JIS K2276.
Lieferumfang: Supplied with sample tube, paddle, unsilvered flask, cork stopper/thermometer support, and an insulated metal case with a viewing slot.