Sampling vessel
Lieferant: STANHOPE SETA
21711-0
STAN21711-0EA 0 EUR
STAN21711-0
Sampling vessel
Probenehmer Probenehmer für Flüssigkeiten
This sampling vessel is designed for petroleum products.
  • Valve can be actuated at any depth
  • Bottom opening valve for non-pressurised use
  • Snatch cord or self actuation when the valve or its extender rod strikes the bottom of the storage tank
Used for sampling applications in the oil and gas industry.
Standards: ASTM D4057/D4306, BS 3195 Part 1, ISO 3170