Hot air tool, HL 1920 E
Lieferant: STEINEL
352103
STEI352103EA 171 EUR
STEI352103
Hot air tool, HL 1920 E
Heizgeräte Heißlufttrockner
A versatile tool for removing and welding sheetings, welding plastics, shrinking on cable sleeves, shaping, soldering, drying, thawing, stripping paint and much more besides. Continuously adjustable from 80 to 600 °C.
  • Ceramic heating element
  • Nine steps temperature setting by thumbwheel
  • High level of quality with optimum weight balance and long life
  • Automatic cut-out protects the tool from overheating
  • Works with 2000 watts of power and airflow control in three settings (150 L/min; 150 to 300 L/min; 300 to 500 L/min)