VZV (gE) Peptide Pool
Lieferant: STEMCELL Technologies
Stimulating T cells with VZV (gE) Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. VZV (gE) Peptide pool is a lyophilised mixture of 153 peptides from the envelope glycoprotein E (gE) of Varicella-zoster virus (VZV; strain Dumas), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 623 on gE. gE is essential for VZV replication (Mo et al., 2002) and may contribute to viral pathogenesis by facilitating epithelial cell-cell contacts (Mo et al., 2000). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').