G418 Disulfat
Lieferant: STRATAGENE
|
Achtung
|
Synonyme:
G-418 Sulfat, G418 Sulfat, Geneticin®, G-418 Disulfat
200398
200399
STRA200398EA
0
EUR
STRA200398
STRA200399
G418 Disulfat
G418 disulphate
Formel:
C₂₀H₄₄N₄O₁₈S₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00058314 CAS-Nummer: 108321-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...