Hygromycin B

Lieferant: STRATAGENE
Gefahr

Synonyme: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol

200715
STRA200715EA 0 EUR
STRA200715
Hygromycin B
Hygromycin B
Formel: C₂₀H₃₇N₃O₁₃
Molecular Weight: 527,5 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06795479
CAS-Nummer: 31282-04-9
EINECS: 250-545-5
UN: 3462
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR