Complete control systems
Lieferant: STRATAGENE
217467 217468
STRA217467EA 166 EUR
STRA217467 STRA217468
Complete control systems
Protein- Expression Protein- Expressionsysteme
The Complete control inducible mammalian expression system is designed to provide the tightest control of expression available in mammalian cells. The system is based on a synthetic ecdysone-inducible receptor and a synthetic receptor recognition element that modulates expression of your gene of interest.
  • Dose-responsive inducible mammalian expression
  • Induction as high as 1700 fold
  • Novel IRES site allows expression of both receptor subunits from the same CMV promoter
  • Retroviral and plasmid gene delivery systems available
Bitte beachten For Research Use Only. Not for Use in Diagnostic Procedures.