Lambda gt11 vector kits
Lieferant: STRATAGENE
234211
STRA234211EA
845
EUR
STRA234211
Lambda gt11 vector kits
Vektoren, Plasmide und Bibliotheken
For use in the construction of cDNA libraries.
- Predigested with EcoR I
- >1×10⁷ pfu/μg lambda DNA must be obtained with test insert (background <1,5%)
These Lambda gt11 vector kits do not allow in vivo excision of cloned fragments. Libraries may be screened with both antibodies and nucleic acid probes.
Bitte beachten For research use only. Not for use in diagnostic procedures.