Lambda genomic vector kits
Lieferant: STRATAGENE
Lambda DASH II and Lambda FIX II vectors offer traditional genomic cloning of 9 to 23 kB fragments.
These vectors are used for genomic library construction contain T3 and T7 RNA promoters for rapid gene walking and high-resolution gene mapping.
Bitte beachten For research use only. Not for use in diagnostic procedures.