MycoSensor qPCR assay kits
Lieferant: STRATAGENE
The MycoSensor PCR and qPCR assay kits detect cultured Mycoplasma infection by real-time qPCR or PCR.
- Detects Mycoplasma infection of cell cultures by qPCR or PCR in less than two hours
- Specific detection of the eight most common Mycoplasma species
- SYBR Green dye detection
The MycoSensor PCR and qPCR assay kits detect Mycoplasma infection of cell cultures by real-time quantitative PCR (qPCR) or PCR in less than two hours. The easy-to-use system can specifically detect the eight most common Mycoplasma species.
The system uses SYBR Green dye detection. The kit includes a primer mix that amplifies the eight most common Mycoplasma species that infect cell cultures. The MycoSensor qPCR assay kit primer mix can detect Mycoplasma infections in cell cultures in less than two hours.