(2,2,6,6-Tetramethyl-3,5-heptandionato-O,O')lithium ≥98%
Lieferant: abcr
AB120843.0005
03-5001-5G
ABCRAB120843.0005EA
0
EUR
ABCRAB120843.0005
STRE03-5001-5G
(2,2,6,6-Tetramethyl-3,5-heptandionato-O,O')lithium ≥98%
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium
Metal TMHD, Volatile Precursors for CVD
Formel:
C₁₁H₁₉LiO₂ Siedepunkt: 295 °C (Zers.) Schmelzpunkt: 265…268 °C |
MDL:
MFCD00142754 CAS-Nummer: 22441-13-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...