4-Methoxyphenylboronsäure ≥97%
Lieferant: Strem Chemicals, Inc.
|
Achtung
|
Synonyme:
(4-Methoxyphenyl)metaborsäure, Dihydroxy(4-methoxyphenyl)boran, 4-Methoxybenzolboronsäure
05-0880-5G
STRE05-0880-5GEA
133
EUR
STRE05-0880-5G
4-Methoxyphenylboronsäure ≥97%
4-Methoxyphenylboronsäure
Boronic Acid and Ester
Formel:
C₇H₉BO₃ Molecular Weight: 151.96 g/mol Schmelzpunkt: 204…206 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039139 CAS-Nummer: 5720-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...