1,3-Bis(1-adamantyl)imidazoliumchlorid ≥97%

Lieferant: Strem Chemicals, Inc.
Achtung

07-0322-250MG
STRE07-0322-250MGEA 242 EUR
STRE07-0322-250MG
1,3-Bis(1-adamantyl)imidazoliumchlorid ≥97%
1,3-Bis(1-adamantyl)imidazoliumchlorid

NHC

Formel: C₂₃H₃₃ClN₂
Schmelzpunkt: 345…346 °C
MDL: MFCD03788919
CAS-Nummer: 131042-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR