1,3-Di-tert-butylimidazoliumchlorid ≥98%

Lieferant: Strem Chemicals, Inc.
Achtung

07-0368-250MG
STRE07-0368-250MGEA 162 EUR
STRE07-0368-250MG
1,3-Di-tert-butylimidazoliumchlorid ≥98%
1,3-Di-tert-butylimidazoliumchlorid

NHC

Formel: C₁₁H₂₁ClN₂
MDL: MFCD03840576
CAS-Nummer: 157197-54-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR