1,3-Diisopropylimidazoliumchlorid ≥97%

Lieferant: Strem Chemicals, Inc.
Achtung

Synonyme: 1,3-Di-i-propylimidazolium chloride, 1,3-Diisopropyl-1H-imidazol-3-ium chloride

07-0485-1G
STRE07-0485-1GEA 142 EUR
STRE07-0485-1G
1,3-Diisopropylimidazoliumchlorid ≥97%
1,3-Diisopropylimidazoliumchlorid

NHC

Formel: C₉H₁₇ClN₂
Molecular Weight: 188.70 g/mol
MDL: MFCD03840577
CAS-Nummer: 139143-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR