1,3-Bis(4-chlorphenyl)imidazoliumchlorid ≥97%

Lieferant: Strem Chemicals, Inc.
Achtung

07-0490-250MG
STRE07-0490-250MGEA 159 EUR
STRE07-0490-250MG
1,3-Bis(4-chlorphenyl)imidazoliumchlorid ≥97%
1,3-Bis(4-chlorphenyl)imidazoliumchlorid

NHC

Formel: C₁₅H₁₁Cl₃N₂
Schmelzpunkt: 251…252 °C
MDL: MFCD03788924
CAS-Nummer: 141556-46-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR