Magnesiumoxid ≥99,99% (Mg-Basis), PURATREM
Lieferant: Strem Chemicals, Inc.
|
Achtung
|
12-2025-5G
STRE12-2025-5GEA
117
EUR
STRE12-2025-5G
Magnesiumoxid ≥99,99% (Mg-Basis), PURATREM
Magnesiumoxid
Formel:
MgO Siedepunkt: 3600 °C Schmelzpunkt: 2800 °C Dichte: 3,58 |
MDL:
MFCD00011109 CAS-Nummer: 1309-48-4 EINECS: 215-171-9 Merck Index: 13,05700 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...