Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')aluminium ≥99% (99.9%-Al)
Lieferant: Strem Chemicals, Inc.
Synonyme:
Al(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)aluminum
13-5000-1G
STRE13-5000-1GEA
0
EUR
STRE13-5000-1G
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')aluminium ≥99% (99.9%-Al)
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')aluminium
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formel:
C₃₃H₅₇AlO₆ Molecular Weight: 576.78 g/mol Siedepunkt: >400 °C/ 150 °C (0.01 mmHg) (Zers./ Subl.) Schmelzpunkt: 255…258 °C |
MDL:
MFCD00058918 CAS-Nummer: 14319-08-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...