(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%

Lieferant: Strem Chemicals, Inc.
Achtung

Synonyme: (S)-(-)-BINAP

15-0151-5G
STRE15-0151-5GEA 724 EUR
STRE15-0151-5G
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

BINAP, Phosphine

Formel: C₄₄H₃₂P₂
Molecular Weight: 622,69 g/mol
Schmelzpunkt: 241…242 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010805
CAS-Nummer: 76189-56-5
Merck Index: 13,01223

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR