(S,S)-(-)-2,3-Bis(diphenylphosphino)butan

Lieferant: Strem Chemicals, Inc.
Achtung

Synonyme: (2S,3S)-(-)-Bis(diphenylphosphino)butane, (2S,3S)-Butane-2,3-diylbis(diphenylphosphine), (S,S)-CHIRAPHOS

15-0160-1G
STRE15-0160-1GEA 690 EUR
STRE15-0160-1G
(S,S)-(-)-2,3-Bis(diphenylphosphino)butan
(S,S)-(-)-2,3-Bis(diphenylphosphino)butan

Phosphine

Formel: C₂₈H₂₈P₂
Molecular Weight: 426.47 g/mol
Schmelzpunkt: 108…109 °C
MDL: MFCD00009836
CAS-Nummer: 64896-28-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR