Trihexyl(tetradecyl)phosphoniumbromid ≥95% (CYPHOS® IL 102)
Lieferant: Strem Chemicals, Inc.
|
Achtung
|
15-6378
STRE15-6378EA
0
EUR
STRE15-6378
Trihexyl(tetradecyl)phosphoniumbromid ≥95% (CYPHOS® IL 102)
Trihexyl(tetradecyl)phosphoniumbromid
CYPHOS IL, Ionic Liquids, Phosphine
Formel:
C₃₂H₆₈BrP Dichte: 0,952 |
MDL:
MFCD04117697 CAS-Nummer: 654057-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...