Trihexyl(tetradecyl)phosphoniumchlorid ≥93% (CYPHOS® IL 101)
Lieferant: Strem Chemicals, Inc.
|
Achtung
|
15-6382
STRE15-6382EA
0
EUR
STRE15-6382
Trihexyl(tetradecyl)phosphoniumchlorid ≥93% (CYPHOS® IL 101)
Trihexyl(tetradecyl)phosphoniumchlorid
CYPHOS IL, Ionic Liquids, Phosphine
Formel:
C₃₂H₆₈ClP Molecular Weight: 519,31 g/mol Dichte: 0,894 |
MDL:
MFCD04112746 CAS-Nummer: 258864-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...