tri-Calciumphosphat (∼37% Ca)

Lieferant: Strem Chemicals, Inc.

20-2039-250G
STRE20-2039-250GEA 38.5 EUR
STRE20-2039-250G
tri-Calciumphosphat (∼37% Ca)
tri-Calciumphosphat
Formel: Ca₃(PO₄)₂
Dichte: 3,14 g/cm³ (20 °C)
MDL: MFCD00015984
CAS-Nummer: 7758-87-4
EINECS: 231-840-8
Merck Index: 12,01741

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR