Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')scandium(III) ≥99% (99.9%-Sc)

Lieferant: Strem Chemicals, Inc.

Synonyme: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)scandium(III), Sc(TMHD)3

21-1000-500MG
STRE21-1000-500MGEA 151 EUR
STRE21-1000-500MG
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')scandium(III) ≥99% (99.9%-Sc)
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')scandium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formel: C₃₃H₅₇O₆Sc
Molecular Weight: 594.758 g/mol
Siedepunkt: 275 °C (Zers.)
Schmelzpunkt: 150…152 °C
MDL: MFCD00169875
CAS-Nummer: 15492-49-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR