1,1'-Bis(diphenylphosphino)ferrocen ≥99%

Lieferant: Strem Chemicals, Inc.
Achtung

Synonyme: DPPF, 1,1-Bis(diphenylphosphino)ferrocene

26-0270-1G
STRE26-0270-1GEA 47.9 EUR
STRE26-0270-1G
1,1'-Bis(diphenylphosphino)ferrocen ≥99%
1,1'-Bis(diphenylphosphino)ferrocen

Phosphine

Formel: C₃₄H₂₈FeP₂
Molecular Weight: 554,39 g/mol
Schmelzpunkt: 180 °C
MDL: MFCD00001422
CAS-Nummer: 12150-46-8
EINECS: 430-420-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR