1,1'-Bis(diisopropylphosphino)ferrocen ≥98%
Lieferant: Strem Chemicals, Inc.
|
Achtung
|
Synonyme:
DiPPF, 1,1'-Bis(di-i-propylphosphino)ferrocene
26-0275-500MG
26-0275-2G
STRE26-0275-500MGEA
73.1
EUR
STRE26-0275-500MG
STRE26-0275-2G
1,1'-Bis(diisopropylphosphino)ferrocen ≥98%
1,1'-Bis(diisopropylphosphino)ferrocen
Phosphine
Formel:
C₂₂H₂₈FeP₂ Molecular Weight: 410.25 g/mol |
MDL:
MFCD02684559 CAS-Nummer: 97239-80-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...