Eisen(III)acetylacetonat ≥99%
Lieferant: Strem Chemicals, Inc.
|
Achtung
|
Synonyme:
Tris(2,4-pentandionato-O,O')eisen(III), Tri(2,4-pentandionato-O,O')eisen(III), Eisen(III)-tris(2,4-pentandionato-O,O'), Eisen(III)-2,4-pentandionat
26-2300-50G
STRE26-2300-50GEA
32.5
EUR
STRE26-2300-50G
Eisen(III)acetylacetonat ≥99%
Eisen(III)acetylacetonat
Formel:
C₁₅H₂₁FeO₆ Molecular Weight: 353.18 g/mol Siedepunkt: 110 °C (2 mmHg) Schmelzpunkt: 184 °C Dichte: 5.24 |
MDL:
MFCD00000020 CAS-Nummer: 14024-18-1 EINECS: 237-853-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...