Pentacarbonyleisen ≥99,5% (99.9+%-Fe)
Lieferant: Strem Chemicals, Inc.
|
Gefahr
|
Synonyme:
Eisenpentacarbonyl
26-2800-250G
STRE26-2800-250GEA
196
EUR
STRE26-2800-250G
Pentacarbonyleisen ≥99,5% (99.9+%-Fe)
Pentacarbonyliron
Metal Carbonyls, Volatile Metal Carbonyls for CVD & ALD, Volatile Precursors for CVD
Formel:
C₅FeO₅ Molecular Weight: 195.9 g/mol Siedepunkt: 103 °C Schmelzpunkt: –20 °C Dichte: 1.490 Flammpunkt: 5 °F |
MDL:
MFCD00011001 CAS-Nummer: 13463-40-6 EINECS: 236-670-8 UN: 1994 ADR: 6.1 I Merck Index: 12,05111 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...