Bis(cyclopentadienyl)nickel ≥99%
Lieferant: Strem Chemicals, Inc.
|
Gefahr
|
Synonyme:
Nickelocene
28-1301-5G
STRE28-1301-5GEA
78.7
EUR
STRE28-1301-5G
Bis(cyclopentadienyl)nickel ≥99%
Bis(cyclopentadienyl)nickel
Metallocenes, Derivatives & Cp Precursors, Volatile Metal Carbonyls for CVD & ALD, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD
Formel:
C₁₀H₁₀Ni Molecular Weight: 188,88 g/mol Schmelzpunkt: 173…174 °C |
MDL:
MFCD00001441 CAS-Nummer: 1271-28-9 EINECS: 215-039-0 UN: 1325 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...