Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')kupfer(II) ≥99%
Lieferant: Strem Chemicals, Inc.
Synonyme:
Cu(TMHD)2, Copper(II)bis(2,2,6,6-tetramethyl-3,5-heptanedionate), Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)
29-3000-1G
STRE29-3000-1GEA
0
EUR
STRE29-3000-1G
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')kupfer(II) ≥99%
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')kupfer(II)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formel:
C₂₂H₃₈CuO₄ Molecular Weight: 430.08 g/mol Siedepunkt: 315 °C/ 88 °C (0.05 mmHg) (Zers./ Subl.) Schmelzpunkt: 198 °C |
MDL:
MFCD00058920 CAS-Nummer: 14040-05-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...