Zirkonium(IV)-tert-butanolat ≥99%
Lieferant: Strem Chemicals, Inc.
|
Achtung
|
Synonyme:
2-Methyl-2-propanol Zirkonium(IV)salz, Zirkonium(IV)-tert-butoxid, Zirkonium(IV)-tert-butylat
40-1749-5G
STRE40-1749-5GEA
210
EUR
STRE40-1749-5G
Zirkonium(IV)-tert-butanolat ≥99%
Zirkonium(IV)-tert-butanolat
Alkoxide, Metal Alkoxides, Volatile Precursors for CVD
Formel:
C₁₆H₃₆O₄Zr Molecular Weight: 383.68 g/mol Siedepunkt: 90 °C (5 mmHg) Dichte: 0.96 Flammpunkt: 185 °F |
MDL:
MFCD00075085 CAS-Nummer: 2081-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...