Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zirkonium(IV) ≥99%

Lieferant: Strem Chemicals, Inc.

40-5000-25G
STRE40-5000-25GEA 618 EUR
STRE40-5000-25G
Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zirkonium(IV) ≥99%
Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zirkonium(IV)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formel: C₄₄H₇₆O₈Zr
Siedepunkt: 180 °C (0,1 mmHg) (Subl.)
Schmelzpunkt: 318…320 °C
MDL: MFCD00145380
CAS-Nummer: 18865-74-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR