Dodecacarbonyltriruthenium ≥99%

Lieferant: Strem Chemicals, Inc.

44-1850-1G
STRE44-1850-1GEA 0 EUR
STRE44-1850-1G
Dodecacarbonyltriruthenium ≥99%
Dodecacarbonyltriruthenium

Metal Carbonyls, Volatile Metal Carbonyls for CVD & ALD, Volatile Precursors for CVD

Formel: C₁₂O₁₂Ru₃
MDL: MFCD00011209
CAS-Nummer: 15243-33-1
EINECS: 239-287-4
UN: 3281
ADR: 6.1,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR