Ruthenium(IV)oxid, wasserfrei ≥99,9% (Ru-Basis)

Lieferant: Strem Chemicals, Inc.

Synonyme: Rutheniumdioxid

44-1977-5G
STRE44-1977-5GEA 487 EUR
STRE44-1977-5G
Ruthenium(IV)oxid, wasserfrei ≥99,9% (Ru-Basis)
Ruthenium(IV)oxid
Formel: O₂Ru
Molecular Weight: 133.07 g/mol
Schmelzpunkt: 1200 Zersetzung
Dichte: 6.97
MDL: MFCD00074857
CAS-Nummer: 12036-10-1
EINECS: 234-840-6
UN: 1479
ADR: 5.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR