Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II)
Lieferant: Strem Chemicals, Inc.
44-6040
STRE44-6040EA
312
EUR
STRE44-6040
Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II)
Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II)
Formel:
C₃₀H₃₆Cl₂N₂P₂Ru |
MDL:
MFCD15144861 CAS-Nummer: 911128-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...