Bis(cyclopentadienyl)ruthenium ≥99% (99.9%-Ru)

Lieferant: Strem Chemicals, Inc.
Achtung

Synonyme: Ruthenocene, Bis(cyclopentadienyl)ruthenium(II)

44-6200-1G
STRE44-6200-1GEA 239 EUR
STRE44-6200-1G
Bis(cyclopentadienyl)ruthenium ≥99% (99.9%-Ru)
Bis(cyclopentadienyl)ruthenium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formel: C₁₀Ru
Molecular Weight: 221,18 g/mol
Schmelzpunkt: 194…198 °C
MDL: MFCD00001442
CAS-Nummer: 1287-13-4
EINECS: 215-065-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR