Chlor(1,5-cyclooctadien)rhodium(I) dimer ≥98%
Lieferant: Strem Chemicals, Inc.
|
Achtung
|
Synonyme:
(1,5-Cyclooctadiene)rhodium chloride dimer
45-0380-5G
STRE45-0380-5GEA
6620
EUR
STRE45-0380-5G
Chlor(1,5-cyclooctadien)rhodium(I) dimer ≥98%
Chlor(1,5-cyclooctadien)rhodium(I) dimer
Formel:
C₁₆H₂₄Cl₂Rh₂ Molecular Weight: 493.08 g/mol |
MDL:
MFCD00012415 CAS-Nummer: 12092-47-6 EINECS: 235-157-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...