Hexadecacarbonylhexarhodium ≥98%
Lieferant: Strem Chemicals, Inc.
45-0820-250MG
AB112997.0250
STRE45-0820-250MGEA
489
EUR
STRE45-0820-250MG
ABCRAB112997.0250
Hexadecacarbonylhexarhodium ≥98%
Hexadecacarbonylhexarhodium
Metal Carbonyls
Formel:
C₁₆O₁₆Rh₆ Dichte: 2,87 |
MDL:
MFCD00011206 CAS-Nummer: 28407-51-4 EINECS: 249-009-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...