[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid-Dichlormethan-Komplex ≥99%

Lieferant: Strem Chemicals, Inc.
Achtung

Synonyme: Dichloro 1,1'-bis(diphenylphosphino)ferrocene palladium (II) dichloromethane, [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) Dichloride Dichloromethane Adduct (1:1), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with dichloromethane, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane complex

46-0450-1G
STRE46-0450-1GEA 112 EUR
STRE46-0450-1G
[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid-Dichlormethan-Komplex ≥99%
[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid-Dichlormethan-Komplex

Metallocenes, Derivatives & Cp Precursors

Formel: C₃₅H₃₀Cl₄FeP₂Pd
Molecular Weight: 816,64 g/mol
MDL: MFCD00792899
CAS-Nummer: 95464-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR