Bariumtrifluormethansulfonat ≥99%

Lieferant: Strem Chemicals, Inc.
Achtung

Synonyme: Barium Bis(trifluoromethanesulphonate), Barium triflate, Barium(II) trifluoromethanesulphonate, Trifluoromethanesulphonic acid Barium(II) salt, Ba(OTf)2, Barium(II) triflate

56-8100-5G
STRE56-8100-5GEA 86.6 EUR
STRE56-8100-5G
Bariumtrifluormethansulfonat ≥99%
Bariumtrifluormethansulfonat

Metal Triflate

Formel: C₂BaF₆O₆S₂
Molecular Weight: 435.45 g/mol
Schmelzpunkt: Zersetzung
MDL: MFCD00143644
CAS-Nummer: 2794-60-7
EINECS: 220-526-6
UN: 1564
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR