Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')terbium(III) ≥99% (99.9%-Tb)
Lieferant: Strem Chemicals, Inc.
Synonyme:
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)terbium(III), Tb(TMHD)3
65-8000-1G
STRE65-8000-1GEA
76.6
EUR
STRE65-8000-1G
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')terbium(III) ≥99% (99.9%-Tb)
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')terbium(III)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formel:
C₃₃H₅₇O₆Tb Molecular Weight: 708.727 g/mol Siedepunkt: 275 °C (Zers.) Schmelzpunkt: 155…156 °C |
MDL:
MFCD00064752 CAS-Nummer: 15492-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...