Decacarbonyldirhenium ≥98%

Lieferant: Strem Chemicals, Inc.

Synonyme: di-Rheniumdecacarbonyl

75-1800-5G
STRE75-1800-5GEA 0 EUR
STRE75-1800-5G
Decacarbonyldirhenium ≥98%
Decacarbonyldirhenium

Metal Carbonyls, Volatile Metal Carbonyls for CVD & ALD, Volatile Precursors for CVD

Formel: C₁₀O₁₀Re₂
Molecular Weight: 652.52 g/mol
Schmelzpunkt: 170 °C (Zers.)
Dichte: 2.87
Lagertemperatur: Raumtemperatur
MDL: MFCD00011198
CAS-Nummer: 14285-68-8
EINECS: 238-202-8
UN: 3466
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR