Dodecacarbonyltetrairidium ≥98%

Lieferant: Strem Chemicals, Inc.

Synonyme: Iridium carbonyl

77-1800-1G
STRE77-1800-1GEA 0 EUR
STRE77-1800-1G
Dodecacarbonyltetrairidium ≥98%
Dodecacarbonyltetrairidium

Metal Carbonyls

Formel: C₁₂Ir₄O₁₂
Molecular Weight: 1104.99 g/mol
Schmelzpunkt: 230 °C (Zers.)
MDL: MFCD00011064
CAS-Nummer: 11065-24-0
UN: 3281
ADR: 6.1 I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR