Platin(II)acetylacetonat ≥98%

Lieferant: Strem Chemicals, Inc.
Achtung

78-1400-5G
STRE78-1400-5GEA 674 EUR
STRE78-1400-5G
Platin(II)acetylacetonat ≥98%
Platin(II)acetylacetonat

Metal Beta-diketonates, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formel: C₁₀H₁₄O₄Pt
Schmelzpunkt: 250…252 °C
Dichte: ∼1 g/cm³ (20 °C)
MDL: MFCD00000028
CAS-Nummer: 15170-57-7
EINECS: 239-223-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR