Vanadylsulfat Hydrat

Lieferant: Strem Chemicals, Inc.
Gefahr

93-2334-25G
STRE93-2334-25GEA 67 EUR
STRE93-2334-25G
Vanadylsulfat Hydrat
Vanadylsulfat Hydrat
Formel: O₅SV·H₂O
MDL: MFCD00149933
CAS-Nummer: 123334-20-3
EINECS: 248-652-7
UN: 2931
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR