Gallium(III)acetylacetonat ≥99,99%, PURATREM

Lieferant: Strem Chemicals, Inc.
Achtung

Synonyme: Tri(2,4-pentandionato-O,O')gallium(III), Gallium(III)-2,4-pentandionat, Tris(2,4-pentandionato-O,O')gallium(III), Gallium(III)-tris(2,4-pentandionato-O,O')

93-3130-25G AB120512.0025 93-3130-5G
STRE93-3130-25GEA 503 EUR
STRE93-3130-25G ABCRAB120512.0025 STRE93-3130-5G
Gallium(III)acetylacetonat ≥99,99%, PURATREM
Gallium(III)acetylacetonat

Metal Beta-diketonates, Volatile Precursors for CVD

Formel: C₁₅H₂₁GaO₆
Molecular Weight: 367.05 g/mol
Siedepunkt: 140 °C (10 mmHg) (Subl.)
Schmelzpunkt: 192…194 °CC >280 °C (Zers.)
Dichte: 1.42
MDL: MFCD00013492
CAS-Nummer: 14405-43-7
EINECS: 238-377-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR