Gallium(III)chlorid, wasserfrei ≥99,999%, körnig, PURATREM

Lieferant: Strem Chemicals, Inc.
Gefahr

93-3140.0025
STRE93-3140.0025EA 255 EUR
STRE93-3140.0025
Gallium(III)chlorid, wasserfrei ≥99,999%, körnig, PURATREM
Gallium(III)chlorid
Formel: Cl₃Ga
Siedepunkt: 201,3 °C
Dichte: 2,47
MDL: MFCD00011018
CAS-Nummer: 13450-90-3
EINECS: 236-610-0
UN: 1759
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR